missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Thienyl)-D-alanine, 95%, Thermo Scientific™
£54.65 - £206.00
Chemical Identifiers
CAS | 62561-76-6 |
---|---|
Molecular Formula | C7H9NO2S |
Molecular Weight (g/mol) | 171.214 |
MDL Number | MFCD00077166 |
InChI Key | WTOFYLAWDLQMBZ-ZCFIWIBFSA-N |
Synonym | 3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712 |
PubChem CID | 7111750 |
ChEBI | CHEBI:77819 |
IUPAC Name | (2R)-2-amino-3-thiophen-2-ylpropanoic acid |
SMILES | C1=CSC(=C1)CC(C(=O)O)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
62561-76-6 | |
171.214 | |
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N | |
7111750 | |
(2R)-2-amino-3-thiophen-2-ylpropanoic acid |
C7H9NO2S | |
MFCD00077166 | |
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712 | |
CHEBI:77819 | |
C1=CSC(=C1)CC(C(=O)O)N |
Specifications
62561-76-6 | |
MFCD00077166 | |
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N | |
(2R)-2-amino-3-thiophen-2-ylpropanoic acid | |
7111750 | |
171.21 | |
Solid |
C7H9NO2S | |
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712 | |
C1=CSC(=C1)CC(C(=O)O)N | |
171.214 | |
CHEBI:77819 | |
95% | |
3-(2-Thienyl)-D-alanine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C