missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole, 99%, Thermo Scientific Chemicals

Product Code. 11490200
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490200 1 g 1g
11400210 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490200 Supplier Thermo Scientific Alfa Aesar Supplier No. H50665.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36770-50-0
Molecular Formula C12H9N5
Molecular Weight (g/mol) 223.239
MDL Number MFCD00107797
InChI Key WOFXUCJTFGKKIF-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5-4-pyridyl-1,2,4-triazole, 2-3-pyridin-4-yl-1h-1,2,4-triazol-5-yl pyridine, 3-2-pyridyl-5-4-pyridyl-1h-1,2,4-triazole, 2-3-4-pyridyl-1h-1,2,4-triazol-5-yl pyridine, 2-5-pyridin-4-yl-1h-1,2,4-triazol-3-yl pyridine, 2-5-4-pyridyl-1h-1,2,4-triazol-3-yl pyridine, acmc-1ah5x, 3-2'-pyridyl-5-4-pyridyl-1,2,4-triazole
PubChem CID 2803172
IUPAC Name 2-(3-pyridin-4-yl-1H-1,2,4-triazol-5-yl)pyridine
SMILES C1=CC=NC(=C1)C2=NC(=NN2)C3=CC=NC=C3

Specifications

Quantity 1 g
Formula Weight 223.24
Percent Purity 99%
Chemical Name or Material 3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.