missing translation for 'onlineSavingsMsg'
Learn More

3-[2-Oxo-3-(trifluoromethyl)-1,2-dihydropyridin-1-yl]propanenitrile, 97%, Thermo Scientific™

Product Code. 10611893
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-60-8
Molecular Formula C9H7F3N2O
Molecular Weight (g/mol) 216.163
MDL Number MFCD00204216
InChI Key CQBBWSPMEJVWQE-UHFFFAOYSA-N
Synonym 3-2-oxo-3-trifluoromethyl pyridin-1 2h-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridone, 3-2-oxo-3-trifluoromethyl-1,2-dihydropyridin-1-yl propanenitrile, 3-2-oxo-3-trifluoromethyl pyridin-1-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyridin-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl-pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridon, 3-2-oxo-3-trifluoromethyl hydropyridyl propanenitrile
PubChem CID 2736731
IUPAC Name 3-[2-oxo-3-(trifluoromethyl)pyridin-1-yl]propanenitrile
SMILES C1=CN(C(=O)C(=C1)C(F)(F)F)CCC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 216.16
Percent Purity 97%
Chemical Name or Material 3-[2-Oxo-3-(trifluoromethyl)-1, 2-dihydropyridin-1-yl]propanenitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'