missing translation for 'onlineSavingsMsg'
Learn More

3-((2-Nitro-4-(trifluoromethyl)-phenyl)amino)propane-1,2-diol, TRC

Product Code. 30009374
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009374 500 mg 500mg
30006719 250 mg 250mg
30008238 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009374 Supplier Toronto Research Chemicals Supplier No. TRCN900998500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104333-00-8
Molecular Formula C10H11N2O4F3
Molecular Weight (g/mol) 280.2
Synonym 3-[[2-Nitro-4-(trifluoromethyl)phenyl]amino]-1,2-propanediol;3-(4-Trifluoromethyl-2-nitroanilino)propane-1,2-diol, HC Yellow 6, HC Yellow No. 6;N-(2,3-Dihydroxypropyl)-2-nitro-4-(trifluoromethyl)aniline;
IUPAC Name 3-[2-nitro-4-(trifluoromethyl)anilino]propane-1,2-diol
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])NCC(CO)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H11F3N2O4/c11-10(12,13)6-1-2-8(9(3-6)15(18)19)14-4-7(17)5-16/h1-3,7,14,16-17H,4-5H2
Formula Weight 280.0671
Chemical Name or Material 3-((2-Nitro-4-(trifluoromethyl)-phenyl)amino)propane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.