missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine, TRC

Product Code. 30476897
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476897 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476897 Supplier Toronto Research Chemicals Supplier No. TRCN378690100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 3-(2-Naphthyl)-L-alanine
CAS 58438-03-2
Molecular Formula C13H13NO2
Quantity 100 mg
InChI Formula InChI=1S/C13H13NO2/c14-12(13(15)16)8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7,12H,8,14H2,(H,15,16)/t12-/m0/s1
SMILES N[C@@H](Cc1ccc2ccccc2c1)C(=O)O
IUPAC Name (2S)-2-amino-3-naphthalen-2-ylpropanoic acid
Molecular Weight (g/mol) 215.25
Formula Weight 215.0946
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.