missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine, 98%, Thermo Scientific™

Product Code. 10604092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10604092 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10604092 Supplier Thermo Scientific Acros Supplier No. 333902500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58438-03-2
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD00066087
InChI Key JPZXHKDZASGCLU-LBPRGKRZSA-N
Synonym 3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid
PubChem CID 185915
IUPAC Name (2S)-2-amino-3-naphthalen-2-ylpropanoic acid
SMILES N[C@@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 215.25
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Naphthyl)-L-alanine
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.