missing translation for 'onlineSavingsMsg'
Learn More

3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester, TRC

Product Code. 30388377
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.50g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388377 250 mg 250mg
30373557 1 g 1g
30369068 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388377 Supplier Toronto Research Chemicals Supplier No. TRCE894935250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester
Analyte or Component Names 3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C19 H24 N2 O6
InChI Formula InChI=1S/C19H24N2O6/c1-5-26-18(23)20(13(3)22)10-9-14-12-21(19(24)27-6-2)17-8-7-15(25-4)11-16(14)17/h7-8,11-12H,5-6,9-10H2,1-4H3
Solution Type Neat
SMILES CCOC(=O)N(CCc1cn(C(=O)OCC)c2ccc(OC)cc12)C(=O)C
IUPAC Name ethyl 3-[2-[acetyl(ethoxycarbonyl)amino]ethyl]-5-methoxyindole-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 376.404
Quantity 250 mg
Formula Weight 376.163
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.