missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester |
| Analyte or Component Names | 3-(2-(N-(Ethoxycarbonyl)acetamido)ethyl)-5-methoxy-1H-indole-1-carboxylic Acid Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 O6 |
| InChI Formula | InChI=1S/C19H24N2O6/c1-5-26-18(23)20(13(3)22)10-9-14-12-21(19(24)27-6-2)17-8-7-15(25-4)11-16(14)17/h7-8,11-12H,5-6,9-10H2,1-4H3 |
| Solution Type | Neat |
| SMILES | CCOC(=O)N(CCc1cn(C(=O)OCC)c2ccc(OC)cc12)C(=O)C |
| IUPAC Name | ethyl 3-[2-[acetyl(ethoxycarbonyl)amino]ethyl]-5-methoxyindole-1-carboxylate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 376.404 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?