missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 10209662
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209662 1 g Amber Glass Bottle 1g
10612045 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209662 Supplier Thermo Scientific Maybridge Supplier No. CC42013DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-08-3
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD07772847
InChI Key KRJWFDTWNMICLY-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methylamine, 3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-yl-ethoxy-benzylamine, 3-2-morpholin-4-yl ethoxy phenyl methylamine, benzenemethanamine, 3-2-4-morpholinyl ethoxy, 1-3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-ylethoxy phenyl methanamine, 3-2-morpholinoethoxy phenylmethylamine, 3-morpholinoethoxy-phenyl methyl amin, 3-2-morpholinoethoxy phenyl methanamine
PubChem CID 7164624
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES C1COCCN1CCOC2=CC=CC(=C2)CN

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 236.31
Percent Purity 95%
Chemical Name or Material [3-(2-Morpholinoethoxy)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.