missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Morpholinoethoxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10015712
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10015712 1 g 1g
10249752 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10015712 Supplier Thermo Scientific Maybridge Supplier No. CC42009DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 857284-07-2
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD07772846
InChI Key DZHYOLUKMGNRTO-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methanol, 3-2-morpholin-4-yl ethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7164622
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES C1COCCN1CCOC2=CC=CC(=C2)CO

Especificaciones

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [3-(2-Morpholinoethoxy)phenyl]methanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.