missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methylphenoxy)pyrrolidine, 95%

Product Code. 11494750
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494750 250 mg 250mg
11404760 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494750 Supplier Thermo Scientific Alfa Aesar Supplier No. H50784.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 46196-54-7 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 46196-54-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD08060984
InChI Key MIVMFYNEGPHLSB-UHFFFAOYSA-N
Synonym 3-2-methylphenoxy pyrrolidine, 3-o-tolyloxy pyrrolidine, 3-o-tolyloxy-pyrrolidine, acmc-1bmts, 3-o-tolyloxy ;pyrrolidine
PubChem CID 18337163
IUPAC Name 3-(2-methylphenoxy)pyrrolidine
SMILES CC1=CC=CC=C1OC2CCNC2

Specifications

Quantity 250 mg
Formula Weight 177.25
Percent Purity 95%
Chemical Name or Material 3-(2-Methylphenoxy)pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.