missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1H-imidazol-1-yl)benzylamine, 97%, Thermo Scientific™

Product Code. p-3527245
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10748825 250 mg Amber Glass Bottle 250mg
10011154 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10748825

Brand: Thermo Scientific Maybridge CC61313CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-62-1
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.25
MDL Number MFCD09065015
InChI Key CECWODAAUDOPCU-UHFFFAOYSA-N
Synonym 3-2-methyl-1h-imidazol-1-yl benzylamine, 3-2-methylimidazol-1-yl phenyl methanamine, 3-2-methyl-1h-imidazol-1-yl phenyl methanamine, 1-3-2-methylimidazol-1-yl phenyl methanamine, 3-2-methylimidazolyl phenyl methylamine, 1-3-2-methyl-1h-imidazol-1-yl phenyl methanamine
PubChem CID 24229639
IUPAC Name [3-(2-methylimidazol-1-yl)phenyl]methanamine
SMILES CC1=NC=CN1C1=CC=CC(CN)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1H-imidazol-1-yl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.