missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1H-imidazol-1-yl)benzylamine, 97%, Thermo Scientific™

Product Code. 10748825
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748825 250 mg Amber Glass Bottle 250mg
10011154 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748825 Supplier Thermo Scientific Maybridge Supplier No. CC61313CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-62-1
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.25
MDL Number MFCD09065015
InChI Key CECWODAAUDOPCU-UHFFFAOYSA-N
Synonym 3-2-methyl-1h-imidazol-1-yl benzylamine, 3-2-methylimidazol-1-yl phenyl methanamine, 3-2-methyl-1h-imidazol-1-yl phenyl methanamine, 1-3-2-methylimidazol-1-yl phenyl methanamine, 3-2-methylimidazolyl phenyl methylamine, 1-3-2-methyl-1h-imidazol-1-yl phenyl methanamine
PubChem CID 24229639
IUPAC Name [3-(2-methylimidazol-1-yl)phenyl]methanamine
SMILES CC1=NC=CN1C1=CC=CC(CN)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1H-imidazol-1-yl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.