missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1H-imidazol-1-yl)benzoic acid, 90%, Thermo Scientific™

Product Code. 10356424
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356424 1 g Amber Glass Bottle 1g
10693045 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356424 Supplier Thermo Scientific Maybridge Supplier No. CC61301DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-59-3
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD09065014
InChI Key ZGRKUYBDLZMGQH-UHFFFAOYSA-N
Synonym 3-2-methyl-1h-imidazol-1-yl benzoic acid, 3-2-methylimidazol-1-yl benzoic acid, 3-2-methylimidazolyl benzoic acid, benzoic acid,3-2-methyl-1h-imidazol-1-yl
PubChem CID 24229638
IUPAC Name 3-(2-methylimidazol-1-yl)benzoic acid
SMILES CC1=NC=CN1C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.21
Percent Purity 90%
Chemical Name or Material 3-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.