missing translation for 'onlineSavingsMsg'
Learn More

3-[(2-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15490796
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449145-16-7
C12H18BNO4S
283.15
MFCD20265185
IZHIWXZQLZLQGQ-UHFFFAOYNA-N
3-2-methyl-1-piperidinyl sulfonyl benzeneboronic acid, 3-2-methylpiperidin-1-ylsulfonyl phenylboronic acid
73995648
[3-(2-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
CC1CCCCN1S(=O)(=O)C1=CC=CC(=C1)B(O)O
This item is not returnable. View return policy
1449145-16-7
C12H18BNO4S
283.15
MFCD20265185
IZHIWXZQLZLQGQ-UHFFFAOYNA-N
3-2-methyl-1-piperidinyl sulfonyl benzeneboronic acid, 3-2-methylpiperidin-1-ylsulfonyl phenylboronic acid
73995648
[3-(2-methylpiperidin-1-yl)sulfonylphenyl]boronic acid
CC1CCCCN1S(=O)(=O)C1=CC=CC(=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 283.15
Percent Purity 97%
Chemical Name or Material 3-[(2-Methyl-1-piperidinyl)sulfonyl]benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'