missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10250545
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10250545 5 g 5g
10304472 250 mg 250mg
10762083 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10250545 Supplier Thermo Scientific Maybridge Supplier No. CC19401DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 28077-41-0
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD02682065
InChI Key YDPHSMPSNLAMJE-UHFFFAOYSA-N
Synonym 3-2-methyl-1,3-thiazol-4-yl benzoic acid, 3-2-methylthiazol-4-yl benzoic acid, benzoic acid,3-2-methyl-4-thiazolyl, 3-2-methyl thiazol-4-yl benzoic acid
PubChem CID 2776495
IUPAC Name 3-(2-methyl-1,3-thiazol-4-yl)benzoic acid
SMILES CC1=NC(=CS1)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1, 3-thiazol-4-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.