missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[(2-hydroxyethyl)amino]propanenitrile, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 3-[(2-hydroxyethyl)amino]propanenitrile |
| CAS | 33759-44-3 |
| Molecular Formula | C5H10N2O |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C5H10N2O/c6-2-1-3-7-4-5-8/h7-8H,1,3-5H2 |
| SMILES | OCCNCCC#N |
| IUPAC Name | 3-(2-hydroxyethylamino)propanenitrile |
| Molecular Weight (g/mol) | 114.15 |
| Formula Weight | 114.0793 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction