missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)propanoic acid, 98%

Product Code. 10344981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10344981

Brand: Thermo Scientific Acros 330100100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 935-13-7 | C7H8O3 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-13-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
InChI Key XLTJXJJMUFDQEZ-UHFFFAOYSA-N
Synonym 3-furan-2-yl propanoic acid, 3-2-furyl propanoic acid, 2-furanpropanoic acid, 2-furanpropionic acid, 3-2-furyl propionic acid, 3-furan-2-yl-propionic acid, furan-2-propionic acid, 3-2-furyl propoinic acid, furfuryl-essigsaure, furfurylacetic acid
PubChem CID 70286
IUPAC Name 3-(furan-2-yl)propanoic acid
SMILES C1=COC(=C1)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56°C to 59°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol,acetone,ethylether,chlorofor
Formula Weight 140.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Furyl)propanoic acid
Show More Show Less

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato