missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Furyl)benzonitrile, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | 3-2-furyl benzonitrile, 3-furan-2-yl benzonitrile, benzonitrile,3-2-furanyl, 3-furan-2-yl-benzonitrile, benzonitrile, 3-2-furanyl, acmc-20mgkx, 3-2-furanyl benzonitrile, 3-2-furyl benzenecarbonitrile, 3-furan-2-yl benzenecarbonitrile |
Formula Weight | 169.18 |
Percent Purity | 97% |
Chemical Name or Material | 3-(2-Furyl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'