missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. p-3175264
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10347124

Brand: Thermo Scientific Maybridge CC46104CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-52-2
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD03990461
InChI Key BTAANNDAXIYWAN-UHFFFAOYSA-N
Synonym 3-2-furyl benzaldehyde, 3-furan-2-yl benzaldehyde, 3-furyl phenyl ketone, benzaldehyde, 3-2-furanyl, ketone, 3-furyl phenyl, 3-furan-2-yl-benzaldehyde, 3-furanylphenylmethanone, 3-furfanylphenylmethanone, 2-3-formylphenyl furan, 3-2-furanyl benzaidehyde
PubChem CID 201640
IUPAC Name 3-(furan-2-yl)benzaldehyde
SMILES O=CC1=CC=CC(=C1)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.