missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)aniline, 97%, Thermo Scientific™

Product Code. 10401504
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401504 5 g 5g
10586314 250 mg 250mg
10786764 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401504 Supplier Thermo Scientific Maybridge Supplier No. CC46114DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 102269-42-1
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD04039071
InChI Key SLNKACMTMZYMNA-UHFFFAOYSA-N
Synonym 3-furan-2-yl aniline, 3-2-furyl aniline, benzenamine,3-2-furanyl, acmc-1c42n, 3-2-furyl aniline hydrochloride
PubChem CID 4161309
IUPAC Name 3-(furan-2-yl)aniline
SMILES C1=CC(=CC(=C1)N)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.