missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)acrylic acid, 99%

Product Code. 11470217
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470217 25 g 25g
11480217 100 g 100g
11329559 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470217 Supplier Thermo Scientific Alfa Aesar Supplier No. B21653.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 539-47-9 | C7H6O3 | 138.12 g/mol

3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

Solubility
Soluble in dichloromethane and diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-47-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003257,MFCD11656544
InChI Key ZCJLOOJRNPHKAV-ONEGZZNKSA-N
Synonym 3-2-furyl acrylic acid, e-3-furan-2-yl acrylic acid, 2-furanacrylic acid, 3-furan-2-yl acrylic acid, furanacrylic acid, 2-furalacetic acid, furacrylic acid, furylacrylic acid, 2e-3-furan-2-yl prop-2-enoic acid, 2-propenoic acid, 3-2-furanyl
PubChem CID 643402
SMILES OC(=O)\C=C\C1=CC=CO1

Specifications

Melting Point 140°C to 144°C
Boiling Point 286°C
Quantity 25 g
Beilstein 81046
Merck Index 14,4297
Solubility Information Soluble in dichloromethane and diethyl ether.
IUPAC Name (E)-3-(furan-2-yl)prop-2-enoic acid
Formula Weight 138.12
Percent Purity 99%
Chemical Name or Material 3-(2-Furyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.