missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol 97+%, Thermo Scientific™

Product Code. 10336614
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10336614 1 g 1g
10634195 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336614 Supplier Thermo Scientific Maybridge Supplier No. CC47009DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-33-8
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.191
InChI Key BYXYWFXMSJTOKB-UHFFFAOYSA-N
Synonym 3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
PubChem CID 18525804
IUPAC Name [5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
SMILES CN1C(=CC(=N1)C2=CC=CO2)CO

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material [3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.