missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Ethylsulfinyl)propyl]-Pentanedioic Acid, TRC

Product Code. 30383994
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383994 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383994 Supplier Toronto Research Chemicals Supplier No. TRCE679805100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[2-(Ethylsulfinyl)propyl]pentanedioic Acid
CAS 116007-24-0
Molecular Formula C10 H18 O5 S
Quantity 100 mg
Synonym 3-[2-(Ethylsulfinyl)propyl]-Pentanedioic Acid, 3-[2-(Ethylsulfinyl)propyl]pentanedioic acid, Pentanedioic acid, 3-[2-(ethylsulfinyl)propyl]-, Clethodim Impurity 3, M17R, M16R
InChI Formula InChI=1S/C10H18O5S/c1-3-16(15)7(2)4-8(5-9(11)12)6-10(13)14/h7-8H,3-6H2,1-2H3,(H,11,12)(H,13,14)
SMILES CCS(=O)C(C)CC(CC(=O)O)CC(=O)O
IUPAC Name 3-(2-ethylsulfinylpropyl)pentanedioic acid
Molecular Weight (g/mol) 250.312
Formula Weight 250.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.