missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Ethoxy-2-oxoethylidene)-4-oxo-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30387947
Change view
Click to view available options
Quantity:
150 mg
Unit Size:
150mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387947 150 mg 150mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387947 Supplier Toronto Research Chemicals Supplier No. TRCE892520150MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Ethoxy-2-oxoethylidene)-4-oxo-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester
CAS 1395498-37-9
Molecular Formula C14H21NO5
Quantity 150 mg
InChI Formula InChI=1S/C14H21NO5/c1-5-19-12(17)8-10-9-15(7-6-11(10)16)13(18)20-14(2,3)4/h8H,5-7,9H2,1-4H3/b10-8+
SMILES CCOC(=O)\C=C\1/CN(CCC1=O)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (3E)-3-(2-ethoxy-2-oxoethylidene)-4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol) 283.32
Formula Weight 283.142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.