missing translation for 'onlineSavingsMsg'
Learn More

3-([[2-[Dihydroxyboranyl)-5-fluorophenyl]methyl]carbamoyl)propanoic Acid, TRC

Product Code. 30399475
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399475 10 mg 10mg
30391211 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399475 Supplier Toronto Research Chemicals Supplier No. TRCH46467010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(((2-(Dihydroxyboranyl)-5-fluorophenyl)methyl)carbamoyl)propanoic Acid
Analyte or Component Names 3-(((2-(Dihydroxyboranyl)-5-fluorophenyl)methyl)carbamoyl)propanoic Acid
Shipping Condition Room Temperature
Molecular Formula C11H13BFNO5
Synonym 3-([[2-[dihydroxyboranyl)-5-fluorophenyl]methyl]carbamoyl)propanoic acid
InChI Formula InChI=1S/C11H13BFNO5/c13-8-1-2-9(12(18)19)7(5-8)6-14-10(15)3-4-11(16)17/h1-2,5,18-19H,3-4,6H2,(H,14,15)(H,16,17)
Solution Type Neat
SMILES FC1=CC(CNC(CCC(O)=O)=O)=C(B(O)O)C=C1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 269.04
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.