missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanoic Acid, Mikromol™
Description
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanoic Acid
Specifications
Specifications
| Chemical Name or Material | 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanoic Acid |
| CAS | 107880-74-0 |
| Analyte or Component Names | 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H12 N4 O2 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Famotidine Imp. F (EP), Famotidine USP Related Compound F, Famotidine USP RC F, Propanoic acid, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, Famotidine Acid Impurity, 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfanyl]-propanoic Acid, Famotidine Related Compound F, Famotidine Impurity F |
| InChI Formula | InChI=1S/C8H12N4O2S2/c9-7(10)12-8-11-5(4-16-8)3-15-2-1-6(13)14/h4H,1-3H2,(H,13,14)(H4,9,10,11,12) |
| SMILES | NC(=Nc1nc(CSCCC(=O)O)cs1)N |
| IUPAC Name | 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?