missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chlorophenyl)propionic acid, 98+%

Product Code. 11348236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348236 1 g 1g
11358236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348236 Supplier Thermo Scientific Alfa Aesar Supplier No. L15306.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1643-28-3 | C9H9ClO2 | 184.619 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-28-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00016547
InChI Key KZMDFTFGWIVSNQ-UHFFFAOYSA-N
Synonym 3-2-chlorophenyl propionic acid, 3-2-chlorophenyl propanoic acid, benzenepropanoic acid, 2-chloro, 2-chlorobenzenepropanoic acid, 2-chloro-benzenepropanoic acid, zlchem 694, pubchem19768, acmc-209dqy, 2-chlorohydrocinnamic acid, ksc494i2t
PubChem CID 95676
IUPAC Name 3-(2-chlorophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CCC(=O)O)Cl

Specifications

Melting Point 94°C to 97°C
Boiling Point 171°C (10 mmHg)
Quantity 1 g
Beilstein 1866789
Formula Weight 184.62
Percent Purity ≥98%
Chemical Name or Material 3-(2-Chlorophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.