missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid, 98%

Product Code. 11392426
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392426 10 g 10g
11302436 50 g 50g
11312436 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392426 Supplier Thermo Scientific Alfa Aesar Supplier No. L06240.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23598-72-3 | C11H8ClNO3 | 237.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23598-72-3
Molecular Formula C11H8ClNO3
Molecular Weight (g/mol) 237.64
MDL Number MFCD00020813
InChI Key UVEPOHNXGXVOJE-UHFFFAOYSA-N
Synonym 3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid
PubChem CID 90203
IUPAC Name 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl

Specifications

Melting Point 187°C to 192°C
Quantity 10 g
Beilstein 791928
Formula Weight 237.64
Percent Purity ≥98%
Chemical Name or Material 3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.