missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Chloromethyl)phenoxy]-N,N-dimethylpropylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10235614
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10235614 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10235614 Supplier Thermo Scientific Maybridge Supplier No. CC59674DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 930111-03-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H19Cl2NO
MDL Number MFCD09702397
Quantity 1 g
Synonym 3-2-chloromethyl phenoxy-n,n-dimethylpropylamine hydrochloride, 2-3-dimethylamino propoxy benzyl chloride hydrochloride, 3-2-chloromethyl phenoxy propyl dimethylamine hydrochloride, 3-2-chloromethyl phenoxy propyl dimethylamine, chloride, 3-2-chloromethyl phenoxy-n,n-dimethylpropan-1-amine hydrochloride, 1-propanamine,3-2-chloromethyl phenoxy-n,n-dimethyl-,hydrochloride 1:1, 3-2-chloromethyl phenoxy-n,n-dimethylpropan-1-amine-hydrogen chloride 1/1
InChI Key FJINIPMBLHIDJV-UHFFFAOYSA-N
SMILES CN(C)CCCOC1=CC=CC=C1CCl.Cl
IUPAC Name 3-[2-(chloromethyl)phenoxy]-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 264.19
PubChem CID 24229614
Formula Weight 264.19
Percent Purity 97%
Chemical Name or Material 3-[2-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.