missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, Mikromol™

Product Code. 17993156
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993156 100 mg 100mg
17943936 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17993156 Supplier Mikromol Supplier No. MM0489.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid
CAS 3919-74-2
Analyte or Component Names 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H7 Cl F N O3
Certifications/Compliance ISO 17025
Synonym Flucloxacillin Sodium Imp. D (EP), Flucloxacillin Imp. D (EP), 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, Flucloxacillin Sodium Impurity D, Flucloxacillin Impurity D
InChI Formula InChI=1S/C11H7ClFNO3/c1-5-8(11(15)16)10(14-17-5)9-6(12)3-2-4-7(9)13/h2-4H,1H3,(H,15,16)
SMILES Cc1onc(c1C(=O)O)c2c(F)cccc2Cl
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol) 255.63
Quantity 100 mg
Formula Weight 255.0098 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.