missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 99%

Product Code. 11312437
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312437 50 g 50g
11302437 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312437 Supplier Thermo Scientific Alfa Aesar Supplier No. L10092.18
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3919-74-2 | C11H7ClFNO3 | 255.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11H7ClFNO3
Molecular Weight (g/mol) 255.63
MDL Number MFCD00055659
InChI Key REIPZLFZMOQHJT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, fcimic acid, 4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid, pubchem8772, acmc-1ckjh, 3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid
PubChem CID 77520
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

Melting Point 201°C to 203°C
Quantity 50 g
Beilstein 806523
Formula Weight 255.63
Percent Purity 99%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.