missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2698969
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10455712

Brand: Thermo Scientific Maybridge SB01884DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11H7ClFNO3
Molecular Weight (g/mol) 255.63
MDL Number MFCD00055659
InChI Key REIPZLFZMOQHJT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, fcimic acid, 4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid, pubchem8772, acmc-1ckjh, 3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid
PubChem CID 77520
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 255.63
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.