missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide (Chlorpromazine Sulfoxide), Mikromol™

Product Code. 30051784
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051784 25 mg 25mg
18002900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051784 Supplier Mikromol Supplier No. MM0420.010025
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide (Chlorpromazine Sulfoxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorpromazine Sulfoxide
CAS 969-99-3
Analyte or Component Names Chlorpromazine Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C17 H19 Cl N2 O S
Certifications/Compliance ISO 17025
Synonym Chlorpromazine Hydrochloride Imp. A (EP), Chlorpromazine Imp. A (EP), 3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Chlorpromazine Sulfoxide, Chlorpromazine Sulphoxide, Chlorpromazine Hydrochloride Impurity A, Chlorpromazine Impurity A
InChI Formula InChI=1S/C17H19ClN2OS/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)22(21)17-9-8-13(18)12-15(17)20/h3-4,6-9,12H,5,10-11H2,1-2H3
SMILES CN(C)CCCN1c2ccccc2S(=O)c3ccc(Cl)cc13
IUPAC Name 3-(2-chloro-5-oxophenothiazin-10-yl)-N,N-dimethylpropan-1-amine
Molecular Weight (g/mol) 334.86
Quantity 25 mg
Formula Weight 334.0907 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.