missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Bromoacetyl)benzonitrile, 95%, Thermo Scientific™

Product Code. 10021984
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021984 25 g Amber Glass Bottle 25g
10722513 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10021984 Supplier Thermo Scientific Maybridge Supplier No. SPB07593EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50916-55-7
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.06
MDL Number MFCD00833267
InChI Key XWCGNFLHRINYCE-UHFFFAOYSA-N
Synonym 3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile
PubChem CID 2735867
IUPAC Name 3-(2-bromoacetyl)benzonitrile
SMILES BrCC(=O)C1=CC=CC(=C1)C#N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula BrCH2C(O)C6H4CN
Quantity 25 g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 3-(2-Bromoacetyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.