missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Boc-amino)ethyl]azetidine, 95%, Thermo Scientific Chemicals

Product Code. 15420689
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162696-31-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD09878605
InChI Key CSPJUZSNJFANNU-UHFFFAOYSA-N
Synonym 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl n-2-azetidin-3-yl ethyl carbamate, 3-2-n-boc-aminoethyl azetidine, tert-butyl 2-3-azetidinyl ethylcarbamate, 2-azetin-3-yl-1-n-boc ethylamine, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester
PubChem CID 37818431
IUPAC Name tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1CNC1

Specifications

Melting Point 84°C to 86°C
Color Brown to White
Formula Weight 200.28
Percent Purity 95%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material 3-[2-(Boc-amino)ethyl]azetidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'