missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£51.80 - £196.00
Chemical Identifiers
CAS | 1257651-17-4 |
---|---|
Molecular Formula | C19H29BN2O5 |
Molecular Weight (g/mol) | 376.26 |
MDL Number | MFCD16294500 |
InChI Key | IYTPRTWBNLJZLK-UHFFFAOYSA-N |
Synonym | 3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate |
PubChem CID | 57415693 |
IUPAC Name | tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate |
SMILES | CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419285
|
Thermo Scientific Alfa Aesar
H51709.03 |
1 g |
£51.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15429285
|
Thermo Scientific Alfa Aesar
H51709.06 |
5 g |
£196.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1257651-17-4 | |
376.26 | |
IYTPRTWBNLJZLK-UHFFFAOYSA-N | |
57415693 | |
CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1 |
C19H29BN2O5 | |
MFCD16294500 | |
3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate | |
tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate |
Specifications
1257651-17-4 | |
MFCD16294500 | |
IYTPRTWBNLJZLK-UHFFFAOYSA-N | |
tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate | |
57415693 | |
97% |
C19H29BN2O5 | |
3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate | |
CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1 | |
376.26 | |
376.26 | |
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only