missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-5-methylindole hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15425016
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15425016 Supplier Thermo Scientific Alfa Aesar Supplier No. H59415.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1010-95-3
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.71
MDL Number MFCD00012683
InChI Key RBHDFGBPJGEYCK-UHFFFAOYSA-N
Synonym 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
PubChem CID 6917145
IUPAC Name hydrogen 2-(5-methyl-1H-indol-3-yl)ethan-1-amine chloride
SMILES [H+].[Cl-].CC1=CC=C2NC=C(CCN)C2=C1

Specifications

Melting Point 290°C to 292°C (decomposition)
Quantity 25 g
Beilstein 3707780
Sensitivity Hygroscopic
Formula Weight 210.71
Percent Purity 97%
Chemical Name or Material 3-(2-Aminoethyl)-5-methylindole hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.