missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-5-bromoindole, 97%

Product Code. 15450459
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15450459

Brand: Thermo Scientific Alfa Aesar H63653.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3610-42-2 | C10H11BrN2 | 239.116 g/mol

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3610-42-2
Molecular Formula C10H11BrN2
Molecular Weight (g/mol) 239.116
MDL Number MFCD00130169
InChI Key CGHUQJRRADEHTQ-UHFFFAOYSA-N
PubChem CID 77158
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN

Specifications

Quantity 1 g
Synonym 5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
Formula Weight 239.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(2-Aminoethyl)-5-bromoindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.