missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-5-bromoindole, 97%, Thermo Scientific Chemicals

£53.10 - £672.00

Chemical Identifiers

CAS 3610-42-2
Molecular Formula C10H11BrN2
Molecular Weight (g/mol) 239.116
MDL Number MFCD00130169
InChI Key CGHUQJRRADEHTQ-UHFFFAOYSA-N
PubChem CID 77158
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15470459
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63653.MD
250 mg
£53.10
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15450459
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63653.03
1 g
£189.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15460459
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63653.06
5 g
£672.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3610-42-2
239.116
CGHUQJRRADEHTQ-UHFFFAOYSA-N
2-(5-bromo-1H-indol-3-yl)ethanamine
C10H11BrN2
MFCD00130169
77158
C1=CC2=C(C=C1Br)C(=CN2)CCN

Specifications

3610-42-2
MFCD00130169
CGHUQJRRADEHTQ-UHFFFAOYSA-N
2-(5-bromo-1H-indol-3-yl)ethanamine
77158
97%
3-(2-Aminoethyl)-5-bromoindole
C10H11BrN2
5-bromotryptamine, 2-5-bromo-1h-indol-3-yl ethanamine, 5-bromo-1h-indole-3-ethylamine, 1h-indole-3-ethanamine,5-bromo, 2-5-bromo-1h-indol-3-yl ethan-1-amine, 1h-indole-3-ethanamine, 5-bromo, 2-5-bromo-1h-indol-3-yl-ethylamine, 5-bromo-tryptamine, 5-br-t, 3-2-aminoethyl-5-bromoindole
C1=CC2=C(C=C1Br)C(=CN2)CCN
239.116
239.11
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 222-779-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only