missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)propionic acid, 96%

Product Code. 15496007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496007 25 g 25g
15476007 1 g 1g
15486007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496007 Supplier Thermo Scientific Alfa Aesar Supplier No. H33210.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 51656-68-9 | C9H8Cl2O2 | 219.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51656-68-9
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD01310807
InChI Key IDEOVPXKPDUXTP-UHFFFAOYSA-N
PubChem CID 2758192
IUPAC Name 3-(2,6-dichlorophenyl)propanoic acid
SMILES C1=CC(=C(C(=C1)Cl)CCC(=O)O)Cl

Specifications

Melting Point 97°C to 99°C
Quantity 25 g
Synonym 3-2,6-dichlorophenyl propanoic acid, 3-2,6-dichlorophenyl propionic acid, 3-2,6-dichlorophenyl propanoicacid, benzenepropanoic acid,2,6-dichloro, benzenepropanoic acid, 2,6-dichloro, 3-2,6-bis chloranyl phenyl propanoic acid
Formula Weight 219.07
Percent Purity 96%
Chemical Name or Material 3-(2,6-Dichlorophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.