missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10178314
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178314 100 g 100g
10551463 10 g 10g
10186762 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10178314 Supplier Thermo Scientific Maybridge Supplier No. SB00572FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3919-76-4
Molecular Formula C11H7Cl2NO3
Molecular Weight (g/mol) 272.08
MDL Number MFCD00051658
InChI Key WQXUUMUOERZZAE-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2',6'-dichlorophenyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-2,6-dichlorophenyl-5-methyl-4-isoxazolecarboxylic acid, pubchem8684, dicloxacillin impurity d, acmc-1cmb3, pyridinium hydroxy propyl sulphobetaine; pps-oh, 3-2,6-dichlorophenyl-5-methylisoxazol-4-carboxylic acid
PubChem CID 77521
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(Cl)C=CC=C1Cl

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 100 g
Formula Weight 272.09
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.