missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride, Thermo Scientific™

Product Code. 11396915
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396915 25 g 25g
10693603 1 g 1g
10435522 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396915 Supplier Thermo Scientific Maybridge Supplier No. SB00576EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4462-55-9
Molecular Formula C11H6Cl3NO2
Molecular Weight (g/mol) 290.524
InChI Key IZQGELJKDARDMZ-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonyl chloride, dcimc chloride, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carbonyl chloride, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonylchloride, 4-isoxazolecarbonyl chloride, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid chloride, 3-2,6-dichlorophenyl-5-methyl isoxazole-4-carbonyl chloride, pubchem8683, dsstox_cid_31536, dsstox_rid_97421
PubChem CID 78212
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 290.53
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.