missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride), Mikromol™
Description
3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride)
Specifications
Specifications
| Chemical Name or Material | 3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride) |
| CAS | 31264-51-4 |
| Analyte or Component Names | 3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 Cl O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-(2,5-Dimethylphenoxy)propyl Chloride, 3-(2,5-Xylyloxy)propyl Chloride, Benzene, 2-(3-chloropropoxy)-1,4-dimethyl- (9CI), Ether, 3-chloropropyl 2,5-xylyl (8CI), 1-Chloro-3-(2,5-dimethylphenoxy)propane |
| InChI Formula | InChI=1S/C11H15ClO/c1-9-4-5-10(2)11(8-9)13-7-3-6-12/h4-5,8H,3,6-7H2,1-2H3 |
| SMILES | Cc1ccc(C)c(OCCCCl)c1 |
| IUPAC Name | 2-(3-chloropropoxy)-1,4-dimethylbenzene |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?