missing translation for 'onlineSavingsMsg'
Learn More

3-(2,5-Dimethoxyphenyl)propionic acid, 96%

Product Code. p-7032193
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15416397

Brand: Thermo Scientific Alfa Aesar H33566.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10538-49-5 | C11H14O4 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10538-49-5
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00060325
InChI Key JENQUCZZZGYHRW-UHFFFAOYSA-N
PubChem CID 66343
IUPAC Name 3-(2,5-dimethoxyphenyl)propanoic acid
SMILES COC1=CC=C(OC)C(CCC(O)=O)=C1

Specifications

Melting Point 61°C to 66°C
Quantity 1 g
Synonym 3-2,5-dimethoxyphenyl propanoic acid, 3-2,5-dimethoxyphenyl propionic acid, benzenepropanoic acid, 2,5-dimethoxy, 2,5-dimethoxyhydrocinnamic acid, 2,5-dimethoxybenzenepropanoic acid, benzenepropanoic acid,2,5-dimethoxy, maybridge1_000808, acmc-1bo6n, ksc503q0f, 2,5-dimethoxy-benzenepropanoic acid
Formula Weight 210.23
Percent Purity 96%
Chemical Name or Material 3-(2,5-Dimethoxyphenyl)propionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.