missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid hydrochloride, 98%, Thermo Scientific™

Product Code. 15480026
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480026 250 mg 250mg
15470026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480026 Supplier Thermo Scientific Alfa Aesar Supplier No. H52689.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957060-89-8
Molecular Formula C13H20BClN2O4
Molecular Weight (g/mol) 314.573
MDL Number MFCD09027255
InChI Key QBUJRGQOXYFCFK-UHFFFAOYSA-N
Synonym 3-2-morpholinoethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-morpholin-4-ylethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethylcarbamoyl phenylboronic acid hcl, 3-2-morpholinoethylcarbamoyl phenylboronic acid, hcl, 3-2-morpholinoethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
PubChem CID 44119393
IUPAC Name [3-(2-morpholin-4-ylethylcarbamoyl)phenyl]boronic acid;hydrochloride
SMILES B(C1=CC(=CC=C1)C(=O)NCCN2CCOCC2)(O)O.Cl

Specifications

Melting Point 190°C to 195°C
Quantity 250 mg
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 314.58
Percent Purity 98%
Chemical Name or Material 3-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.