missing translation for 'onlineSavingsMsg'
Learn More

3-[2-[4-[(EZ)-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]phenyl](hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Mikromol

Product Code. 30464221
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30464221 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464221 Supplier Mikromol Supplier No. MM0491.450025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[2-[4-[(EZ)-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]phenyl](hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Risperidone EP Impurity J
CAS 1346606-24-3
Analyte or Component Names Risperidone EP Impurity J
Shipping Condition Room Temperature
Molecular Formula C35 H40 F2 N6 O3
Certifications/Compliance ISO 9001
Synonym 3-[2-[4-[(Z)-(4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl)phenyl](hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, (Risperidone Impurity, Mixture of E/Z isomers)
InChI Formula InChI=1S/C35H40F2N6O3/c1-22-27(35(44)43-14-3-2-4-32(43)38-22)13-17-41-15-9-23(10-16-41)33(39-45)28-7-5-25(36)20-30(28)42-18-11-24(12-19-42)34-29-8-6-26(37)21-31(29)46-40-34/h5-8,20-21,23-24,45H,2-4,9-19H2,1H3
Solution Type Neat
SMILES CC1=C(CCN2CCC(CC2)C(=NO)c3ccc(F)cc3N4CCC(CC4)c5noc6cc(F)ccc56)C(=O)N7CCCCC7=N1
IUPAC Name 3-[2-[4-[C-[4-fluoro-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]phenyl]-N-hydroxycarbonimidoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol) 630.7273
Quantity 25 mg
Formula Weight 630.313 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.