missing translation for 'onlineSavingsMsg'
Learn More

3-[2-[4-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]benzolyl]piperidin-1-yl]ethyl-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, TRC

Product Code. 30399389
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399389 25 mg -
30405458 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399389 Supplier Toronto Research Chemicals Supplier No. TRCF59159025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329796-66-8
Molecular Formula C35 H39 F2 N5 O3
Molecular Weight (g/mol) 615.71
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-[4-fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]benzoyl]-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-, 3-[2-[4-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]benzoyl]-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]benzoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido-[1,2-a]pyrimidin-4-one, Risperidone Imp. I (EP)
IUPAC Name 3-[2-[4-[4-fluoro-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]benzoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3N4CCC(CC4)c5noc6cc(F)ccc56)C(=O)N7CCCCC7=N1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C35H39F2N5O3/c1-22-27(35(44)42-14-3-2-4-32(42)38-22)13-17-40-15-9-24(10-16-40)34(43)28-7-5-25(36)20-30(28)41-18-11-23(12-19-41)33-29-8-6-26(37)21-31(29)45-39-33/h5-8,20-21,23-24H,2-4,9-19H2,1H3
Formula Weight 615.3021
Chemical Name or Material 3-[2-[4-[4-Fluoro-2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]benzoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido-[1,2-a]pyrimidin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.