missing translation for 'onlineSavingsMsg'
Learn More

3-[(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)oxy]propanoic Acid, TRC

Product Code. 30386547
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386547 500 mg
30383597 100 mg
30388285 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386547 Supplier Toronto Research Chemicals Supplier No. TRCD483670500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926268-51-1
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.26
IUPAC Name 3-[(2,2-dimethyl-3H-1-benzofuran-7-yl)oxy]propanoic acid
SMILES CC1(C)Cc2cccc(OCCC(=O)O)c2O1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H16O4/c1-13(2)8-9-4-3-5-10(12(9)17-13)16-7-6-11(14)15/h3-5H,6-8H2,1-2H3,(H,14,15)
Formula Weight 236.1049
Chemical Name or Material 3-[(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)oxy]propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.