missing translation for 'onlineSavingsMsg'
Learn More

3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride, TRC

Product Code. 30381526
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381526 500 mg
30378712 250 mg
30384918 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381526 Supplier Toronto Research Chemicals Supplier No. TRCD435678500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52314-67-7
Molecular Formula C8H9Cl3O
Molecular Weight (g/mol) 227.52
Synonym 3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichloroethenyl)cyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarbonyl Chloride, 3-(2,2-Dichlorovinyl)-2,2-dimethyl-1-cyclopropanecarbonyl Chloride
IUPAC Name 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carbonyl chloride
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H9Cl3O/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3
Formula Weight 225.9719
Chemical Name or Material 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.